Anti-ID3 Mouse Monoclonal Antibody [clone: OTI7B6]
Catalog # ORIGTA500811
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor of differentiation 3
- Antigen symbol:ID3
- Clonality:Monoclonal
- Clone:OTI7B6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:13.0 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: ID3 (inhibitor of DNA binding 3, dominant negative helix-loop-helix protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human