Anti-SCN10A Mouse Monoclonal Antibody [clone: S134-12]
ORIGTA309443
: OriGene
- Antibody type:Primary
- Antigen name:sodium channel, voltage-gated, type X, alpha subunit
- Antigen symbol:SCN10A
- Clonality:Monoclonal
- Clone:S134-12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Recommended Dilutions:
Type: Primary
Antigen: SCN10A (sodium channel, voltage-gated, type X, alpha subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat