Order Entry
Finland
ContactUsLinkComponent
Anti-SCN8A Mouse Monoclonal Antibody [clone: S87A-10]
Anti-SCN8A Mouse Monoclonal Antibody [clone: S87A-10]
Catalog # ORIGTA309441
Supplier:  OriGene
Anti-SCN8A Mouse Monoclonal Antibody [clone: S87A-10]
Catalog # ORIGTA309441
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, voltage gated, type VIII, alpha subunit
  • Antigen symbol:
    SCN8A
  • Clonality:
    Monoclonal
  • Clone:
    S87A-10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    459 to 476
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions:

Type: Primary
Antigen: SCN8A (sodium channel, voltage gated, type VIII, alpha subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat