Order Entry
Finland
ContactUsLinkComponent
Anti-IFNG Rat Monoclonal Antibody [clone: XMG1.2]
Anti-IFNG Rat Monoclonal Antibody [clone: XMG1.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IFNG Rat Monoclonal Antibody [clone: XMG1.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 40-7311-U100
  • 40-7311-U500
  • Antigen symbol:
  • IFNG
  • IFNG
  • Antigen name:
  • Interferon gamma
  • Interferon gamma
  • Antigen synonyms:
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • XMG1.2
  • XMG1.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 15978
  • 15978
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Functional Assays, Immunofluorescence Microscopy
  • Functional Assays, Immunofluorescence Microscopy

Specifications

Related Information