Order Entry
Finland
ContactUsLinkComponent
Anti-CAMK1D (CAMK1 delta) Rabbit Polyclonal Antibody
Anti-CAMK1D (CAMK1 delta) Rabbit Polyclonal Antibody
Catalog # ABGEAP7204A
Supplier:  Abgent
undefined
Anti-CAMK1D (CAMK1 delta) Rabbit Polyclonal Antibody
Catalog # ABGEAP7204A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium/calmodulin-dependent protein kinase type 1D
  • Antigen symbol:
    CAMK1D
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal (317–333 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    CaM-KI delta|CaM kinase I delta|CKLiK|CamKI-like protein kinase|Camk1D|CaM kinase ID|CaMKI delta|CaMKI-delta|CAMK1D
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CAMK1D (CAMK1 delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human