Anti-CDCA3 Rabbit Polyclonal Antibody
ABGEAP6614A
: Abgent
- Antibody type:Primary
- Antigen name:Cell division cycle associated 3
- Antigen symbol:CDCA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CDCA3|GRCC8|TOME-1|C8|Cell Division Cycle-associated protein 3|Gene-rich cluster protein C8|TOME1|Trigger of mitotic entry protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CDCA3 (cell division cycle associated 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse