Anti-AAGAB Rabbit Polyclonal Antibody
Catalog # ABGEAP5483B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Alpha- and Gamma-Adaptin Binding Protein
- Antigen symbol:AAGAB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:AAGAB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: AAGAB (alpha- and gamma-adaptin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human