Order Entry
Finland
ContactUsLinkComponent
Anti-Semaphorin 5A Rabbit Polyclonal Antibody
Anti-Semaphorin 5A Rabbit Polyclonal Antibody
Catalog # ABGEAP2712C
Supplier:  Abgent
undefined
Anti-Semaphorin 5A Rabbit Polyclonal Antibody
Catalog # ABGEAP2712C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Semaphorin 5A
  • Antigen symbol:
    SEMA5A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    Semaphorin-F|SEMAF|SEMA5A|semF|Semaphorin-5A|Semaf|Sema F
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: Semaphorin 5A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse