Anti-Serine Palmitoyltransferase (SPTLC2) Rabbit Polyclonal Antibody
ABGEAP2533B
: Abgent
- Antibody type:Primary
- Antigen name:Serine Palmitoyltransferase
- Antigen symbol:SPT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Serine-palmitoyl-CoA transferase 2|SPT2|LCB 2|Serine palmitoyltransferase 2|Long chain base biosynthesis protein 2|SPT 2|SPTLC2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: Serine Palmitoyltransferase (SPTLC2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse