Anti-ZNF417 Rabbit Polyclonal Antibody
ABGEAP12356C
: Abgent
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 417
- Antigen symbol:ZNF417
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:KIAA1139|ANKS5B|Caskin-2|CASK-interacting protein 2|CASKIN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ZNF417 (zinc finger protein 417)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human