Anti-IQCC Rabbit Polyclonal Antibody
AVIVARP70593_P050
- Antibody type:Primary
- Antigen name:IQ motif containing C
- Antigen symbol:IQCC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:55721
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: IQCC (IQ motif containing C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human