Anti-TPRA1 Rabbit Polyclonal Antibody
AVIVARP69268_P050
- Antibody type:Primary
- Antigen name:transmembrane protein, adipocyte asscociated 1
- Antigen symbol:TPRA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:131601
- Antigen synonyms:GPR175|TMEM227|TPRA40
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TPRA1 (transmembrane protein, adipocyte asscociated 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human