Anti-RBMX Rabbit Polyclonal Antibody
Catalog # AVIVARP61803_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:RNA binding motif protein, X-linked
- Antigen symbol:RBMX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:27316
- Antigen synonyms:RBMXRT|RBMXP1|HNRNPG|HNRPG|RNMX|hnRNP-G
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RBMX (RNA binding motif protein, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human