Anti-TRPC4AP Rabbit Polyclonal Antibody
AVIVARP55313_P050
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily C, member 4 associated protein
- Antigen symbol:TRPC4AP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26133
- Antigen synonyms:C20orf188|TRUSS|TRRP4AP|PPP1R158
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TRPC4AP (transient receptor potential cation channel, subfamily C, member 4 associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: