Anti-SLC24A4 Rabbit Polyclonal Antibody
Catalog # AVIVARP44134_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 24 (sodium/potassium/calcium exchanger), member 4
- Antigen symbol:Solute carrier family 24, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:123041
- Antigen synonyms:AI2A5|SHEP6|NCKX4|SLC24A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC24A4 (solute carrier family 24 (sodium/potassium/calcium exchanger), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: