Anti-SLC9A3R2 Rabbit Polyclonal Antibody
AVIVARP43890_P050
- Antibody type:Primary
- Antigen name:solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2
- Antigen symbol:Solute carrier family 9, member 3R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:65962
- Antigen synonyms:Nherf2|NHERF-2|E3karp|2010007A20Rik|Tka-1|0610011L07Rik|Sip1|1200011K07Rik|Sryip1|Octs2|Sip-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC9A3R2 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat