Anti-SLC13A3 Rabbit Polyclonal Antibody
Catalog # AVIVARP41438_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
- Antigen symbol:Solute carrier family 13, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:64849
- Antigen synonyms:SDCT2|NADC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC13A3 (solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: