Anti-SCN3B Rabbit Polyclonal Antibody
AVIVARP37699_P050
- Antibody type:Primary
- Antigen name:sodium channel, voltage gated, type III beta subunit
- Antigen symbol:SCN3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55800
- Antigen synonyms:BRGDA7|ATFB16|SCNB3|HSA243396
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SCN3B (sodium channel, voltage-gated, type III, beta subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: