Anti-AHR Rabbit Polyclonal Antibody
AVIVARP31635_T100
- Antibody type:Primary
- Antigen name:aryl hydrocarbon receptor
- Antigen symbol:AHR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:196
- Antigen synonyms:bHLHe76
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: AHR (aryl hydrocarbon receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: