Anti-SLC4A4 Rabbit Polyclonal Antibody
BOSSBS-2096R
: Bioss
- Antibody type:Primary
- Antigen name:Solute Carrier Family 4 (sodium bicarbonate cotransporter), Member 4
- Antigen symbol:Solute carrier family 4, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:NBCe1-A|KNBC|hhNMC|NBC2|SLC4A5|HNBC1|pNBC|NBC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: SLC4A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat