Polyvinyl alcohol 5-88, EMPROVE® EXPERT Ph. Eur., USP, JPE suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
Melting Pt:160…240 °C
Storage Temperature:Ambient