Carboys with spigot, SP Bel-Art
BELAF118470010
- Capacity:4 L
- Description:Carboy with spigot, PP cap
- Material:LDPE
- Pk:1
Translucent carboys made of lightweight, durable polyethylene with extra-heavy walls for long life and dependability.
- Lighter than glass and easy to handle
- Heavy knurling on cap provides excellent grip for opening and closing
- Resistant to corrosive acids and alkalis
- The spigot requires only half turn to open or close
- Smooth sides accept adhesive labels and can be marked with most indelible markers
Ideal for dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.