Dishes, crystallising, quartz
Supplier: QUARTZTECHNISCHE WERKSTATTEN
New Product
Certificates
About this item
Quartz glass, clear.
- Extremely low expansion
- Best possible elastricity and thermal shock resistance
- High transformation and softening point temperature
- Low heat conductivity
- Low electrical conductivity
For a wide range of applications.