Order Entry
Finland
ContactUsLinkComponent
Jerricans, Narrow Neck, HDPE, Natural
Jerricans, Narrow Neck, HDPE, Natural
Catalog # RIXI6010010045WC
Supplier:  RIXIUS
New Product
undefined
Jerricans, Narrow Neck, HDPE, Natural
Catalog # RIXI6010010045WC
Supplier:  RIXIUS
New Product

Specifications

  • Thread:
    45 mm
  • Capacity:
    10 L
  • Colour:
    Natural
  • Description:
    Jerrycan with screwcap
  • Seq:
    03
  • W×D×H:
    193×234×298 mm

Specifications

About this item

Universal canister, made of HDPE, natural, for industry, trade and laboratories. Ideal for packaging, storage and transportation.

  • Good chemical resistance
  • Good temperature resistance
  • With scale
  • Screw cap must be ordered seperately