Electrodeless discharge lamps
Supplier: PerkinElmer
Certificates
About this item
These electrodeless discharge lamps are preferred for certain volatile elements. They offer better precision and lower detection limits for analyses that are 'noisy' due to weak hollow cathode emission. PerkinElmer system 2 EDLs consist of the element or a salt of the element sealed in a quartz bulb containing an inert gas atmosphere. When an RF field of sufficient power is applied, the inert gas is ionised and the coupled energy vaporizes the element and excites the atoms inside the bulb, resulting in the emission of the characteristic spectrum.
- Interchangeable lamp sleeves
- Dual-channel power supply with RF driver assemblies
- Lamp sleeve with pre-aligned bulb for the element of interest