Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Soap Dispensers and Lotion Soap Dispensers
Accessories for GOJO and PURELL Dispensers
Accessories for GOJO and PURELL Dispensers
Accessories for GOJO and PURELL Dispensers
Catalog # GOJO1045-WHT-12
Supplier:
GOJO Industries - Europe Ltd
Expand image
Accessories for GOJO and PURELL Dispensers
Catalog # GOJO1045-WHT-12
Supplier:
GOJO Industries - Europe Ltd
Specifications
Colour:
White
Description:
Floor and wall protection shield for any LTX™ or ADX™ dispensers
Seq:
209
Pk:
1
Specifications
About this item
Product Details
Product Details