Amino acid F reagent
HACH2386442
: Hach
:
- Pk:100 mL
- Description:Amino acid F reagent solution for Heteropoly Blue Method
- Range:3 - 1000 µg/litre SiO₂
Reagent for the determination of ultra low range silica by the heteropoly blue method. Hach method range: 3 to 1000 µg/L SiO₂.