Order Entry
Finland
ContactUsLinkComponent
 
 
 
 
Octyl phenol ethoxylate, BAKER ANALYZED®, J.T.Baker®
  X198-05
 :  Avantor
 :  
undefined
 
 
 
Octyl phenol ethoxylate, BAKER ANALYZED®, J.T.Baker®
  X198-05
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    120 mL
  • Seq:
    2470

 

 

  • For Laboratory,Research,or Manufacturing Use:
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤ 125
  • Specific Gravity at 25°/25°C:
    1.055 - 1.070

 

 

 :
 :3082
 :9,III
 :GHS05
 :GHS07
 :GHS09
 :9036-19-5
 :MFCD00128254