qPCR master mixes, Fast EvaGreen®
Supplier: Biotium
Certificates
About this item
A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.
- 2X qPCR master mix for fast cycling protocols
- EvaGreen® dye-based
- Contains Cheetah™ Taq hotstart DNA polymerase
Fast Evagreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.
Fast Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.
Specifications
- Application:qPCR dye
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Classified as non-hazardous to aquatic life under CCR title 22 regulations