Order Entry
Finland
ContactUsLinkComponent
Racks for upright freezers
Racks for upright freezers
Catalog # HEATHS28622
Supplier:  HEATHROW
undefined
Racks for upright freezers
Catalog # HEATHS28622
Supplier:  HEATHROW

Specifications

  • Compartments:
    6 (2×3)
  • W×D×H ext.:
    459×159×271 mm
  • Pk:
    1
  • Cryobox height:
    133 mm

Specifications

About this item

Stainless steel.

For upright freezers and used for 15 and 50 ml tube box storage. Corrosion-resistant construction with (2×3) rack spaces.