Covers for 4" chip trays, H44
Supplier: ENTEGRIS
Certificates
About this item
Tray covers in various standard materials are shown below.
- STAT-PRO® 150 covers and trays are not recommended for use with devices less than .012"
- STAT-PRO® 150 material covers are recommended for use only with STAT-PRO® 150 material trays
Articles may not be available in all countries. Please contact your local Avantor office.