Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 0.9% in water normal saline solution
Sodium chloride 0.9% in water normal saline solution
Sodium chloride 0.9% in water normal saline solution
Supplier:
G-Biosciences
Certificates
Expand image
Sodium chloride 0.9% in water normal saline solution
Supplier:
G-Biosciences
Certificates
About this item
CAS Number:
7647-14-5
:
Formula:
NaCl
MDL Number:
MFCD00003477
:
58.44 g/mol
Storage Temperature:
Ambient
EINECS:
231-598-3
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
GENO786-561_TDS_EN
Documentation