Order Entry
Finland
ContactUsLinkComponent
InGel™ Blue for protein digestion
InGel™ Blue for protein digestion
Catalog # 786-681
Supplier:  G-Biosciences
CAS Number:  
undefined
InGel™ Blue for protein digestion
Catalog # 786-681
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    InGel™ Blue, 100 spots
Specifications
About this item
InGel™ Blue provides a complete set of reagents for the in-gel tryptic digestion and extraction of peptides for mass spectrometry (MALDI and LC MS/MS). The kit is specifically designed for use with Coomassie® or fluorescent stained protein spots/bands.
  • For the in-gel tryptic digestion of proteins
  • Compatible with Coomassie® and fluorescent stained proteins
  • Supplied with mass spectrometry grade trypsin
  • Supplied with destaining, reducing, alklating and peptide extraction reagents