Cleaning solutions/spray for electrophoresis gel plates, GELPLATE-clean™
Supplier: G-Biosciences
Certificates
About this item
GELPLATE-clean™ removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. It is suitable for sequencing and protein electrophoresis gel plates.
- Prevent poor electrophoretic band migration
- Prevent band distortions
- Prevent poor image development
Delivery: Supplied as 2 solutions in 2× 250 ml spray bottles. Refill solutions are available and are offered separately.