Order Entry
Finland
ContactUsLinkComponent
CytoScan™ LDH Cytotoxicity Assay kit
CytoScan™ LDH Cytotoxicity Assay kit
Catalog # 786-210
Supplier:  G-Biosciences
CAS Number:  
undefined
CytoScan™ LDH Cytotoxicity Assay kit
Catalog # 786-210
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1.000 Assays
  • Description:
    CytoScan™-LDH Cytotoxicity Kit
  • Tests per kit:
    1000 assays
Specifications
About this item
CytoScan™ LDH Cytotoxicity assay quantitatively measures the cytosolic, lactate dehydrogenase (LDH) enzyme, which is released from damaged cells. The released LDH is measured with a coupled enzymatic reaction that results in the conversion of a tetrazolium salt (iodonitrotetrazolium; (INT)) into a red color formazan by diaphorase. The resulting formazan is measured at 490 nm.
  • Colorimetric assay
  • Quantitatively measures LDH release
  • For the detection of cell toxicity, death, viability or proliferation
  • Ideal for high throughput screening