Order Entry
Finland
Orders LinkContactUsLinkComponent
Ribbons for label printer BBP™33

Ribbons for label printer BBP™33

Ribbon B30-R6400, 60,9 m, Black
Catalog # BRDY143118
Supplier:   Brady
Ribbons for label printer BBP™33
Catalog # BRDY143118
Supplier:  Brady
Ribbons for label printer BBP™33
Ribbons for label printer BBP™33

Ribbon B30-R6400, 60,9 m, Black

Price
106,00 €
Log in for availability
1 * 1 Items
Product Details & Documents

'Drop & Click' print ribbons for quick and easy supply changes.

  • Tough, smear-proof ribbons for long-lasting printed text
  • Print can withstand years in industrial areas and outdoor conditions
  • No calibrating, no threading, no waste, no hassle

Width: 104,39 mm

Specifications
  • Colour:
    Black
  • Description:
    Ribbon B30-R6400
  • Length:
    60,9 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.