Order Entry
Finland
ContactUsLinkComponent
Immobilised Ficin
Immobilised Ficin
Catalog # 786-793
Supplier:  G-Biosciences
CAS Number:  
undefined
Immobilised Ficin
Catalog # 786-793
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Ficin, immobilised
Specifications
About this item
Ficin (or Ficain) is a cysteine protease enzyme (EC 3.4.22.3) isolated from fig latex is that has the endopeptidase activity to cleave Immunoglobulin G (IgG) molecules in the hinge region.
  • Generate Fab and F(ab‘)₂ fragments
  • For digestion of mouse IgG₁
  • Eliminates contamination by Ficin