One-piece hex-head fittings
Supplier: VICI JOUR RESEARCH
Certificates
About this item
One-piece hex-head fittings made from PEEK are easy to use with the ValvTool and the Color-It Color-Coded Fingertight adapters. The 10-32 thread is suitable for standard high-pressure female connections. They are available both in a short and long version.
- Ideal for use with PEEK tubing
- Withstand pressures up to 350 bar (5000 psi)
- For any high pressure fittings