Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ponceau S
Ponceau S 0.1% in dil. acetic acid membrane stain
Ponceau S 0.1% in dil. acetic acid membrane stain
Ponceau S 0.1% in dil. acetic acid membrane stain
Supplier:
G-Biosciences
Certificates
Expand image
Ponceau S 0.1% in dil. acetic acid membrane stain
Supplier:
G-Biosciences
About this item
CAS Number:
6226-79-5
:
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:
MFCD00003892
:
760.56 g/mol
Storage Temperature:
Ambient
Related Information
Documentation
Manual
786-575_protocol.pdf
786-576_protocol.pdf
Documentation