Capillary columns, Rxi®-5HT (fused silica)
: Restek
Fused capillary columns for high-temperature applications such as mineral oil extraction and analysis.
Low polarity phase; diphenyl dimethyl polysiloxane
Temperature range: –60 to +400 °C
Similar phases: DB-5ht, VF-5ht, ZB-5HTinferno.