Cooling bags, Fold’N Cool™
Supplier: CAMPING GAZ
Certificates
About this item
The Fold'N Cool™ soft foldable coolers are comfortable to carry and are available in various sizes and could be folded to store after use.
- Adjustable shoulder strap
- Collapsible cooler
- EPE closed cells foam 6 mm
The bags with 10 litre capacity are light, which holds 8×0,5 litre bottles vertically and 10×0,5 litre bottles horizontally. The bags with 30 litre capacity can hold 9×1,5 litre bottles vertically and 12×1,5 litre bottles horizontally. The bags with 20 litre capacity can hold 6×1,5 litre bottles vertically and 5×1,5 litre bottles horizontally. The bags with 5 litre capacity can hold 6×0,33 litre bottles vertically and horizontally.