Order Entry
Finland
ContactUsLinkComponent
Centrifuge tubes
Centrifuge tubes
Catalog # BRND778013
Supplier:  Brand
undefined
Centrifuge tubes
Catalog # BRND778013
Supplier:  Brand

Specifications

  • Closure type:
    Without cap
  • Ø×L:
    17×113 mm
  • Capacity:
    15 ml
  • Seq:
    01
  • Pk:
    100

Specifications

About this item

Duran® borosilicate glass.

  • Withstands forces up to 3500×g

Conical bottom, beaded rim, ungraduated.