Order Entry
Finland
ContactUsLinkComponent
Eyewash station, HypaClens
Eyewash station, HypaClens
  118-0283
undefined
Eyewash station, HypaClens
  118-0283

 

  • Description:
    Eyewash station with 2×500 ml eyewash bottles
  • Pk:
    1

 

 

This wall-mountable eyewash station holds two 500 ml HypaClens eyewash bottles.

  • Integrated mirror allows for self-administration of eyewash
  • Compact design of the eyewash station makes it ideal for any work place with low risks

W×D×H (mm): 250×60×300

 : Station is supplied with two bottles of eyewash solution. Wall fixings are not included. Refills are available separately, please contact your local Avantor sales office for further information.