Disposable weighing boats, Sterilin®
Supplier: Sterilin
Certificates
About this item
Polystyrene, white.
- Radiused corners and sloping sides for complete sample transfer
- Flat base for a secure firm rest on the balance pan
- Suitable for coloured powders and samples
- Diamond shape is particulrly flexible to allow the formation of a funnel for easy pouring to another vessel
Manufactured from high impact polystyrene with a hydrophobic surface that ensures no absorption of water from atmosphere or sample. The anti-staic version is made from PS material, modified to significantly reduce the charge relaxation time in comparison to standard weigh boats - the majority of electrostatic charge dissipates within seconds when earthed on the balance pan