Büchner vacuum filtration funnels
Supplier: SciLabware
Certificates
About this item
Borosilicate glass 3.3
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
Büchner vacuum filtration funnel with glass sintered disc. It is designed with ground glass cone to allow direct filtration into Büchner flasks.
Ordering information: Büchner flasks must be ordered separately.