Dropping funnels, cylindrical, Quickfit®
Supplier: SciLabware
Certificates
About this item
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: Complies with ISO 4800