Order Entry
Finland
ContactUsLinkComponent
pH/mV/°C/rH meter, handheld, GMH 3531
pH/mV/°C/rH meter, handheld, GMH 3531
Catalog # GRSG602076
Supplier:  Greisinger
undefined
pH/mV/°C/rH meter, handheld, GMH 3531
Catalog # GRSG602076
Supplier:  Greisinger

Specifications

  • Temperature range:
    −5,0...+150,0 °C
  • Description:
    Handheld pH/mV/°C/rH meter, GMH 3531
  • Model:
    GMH 3531
  • Temperature accuracy:
    ±0,2 °C (−5...+100 °C), otherwise ±0,4
  • Temperature resolution:
    0,1 °C
  • Connections:
    BNC (pH, Redox, rH), 2 × 4 mm banana socket for Pt1000, 2-wire
  • Display:
    LCD (two lines, 2×4 digits, 12,4 + 7 mm height)
  • Outputs:
    Serial interface (3,5 mm jack); analog output
  • Power supply:
    9 V battery, additional socket for external 10,5 - 12 V direct current power supply
  • Temperature compensation:
    Automatic (ATC) in the operating modes "pH" if temperature probe is used (0...105 °C); otherwise manual input
  • Weight:
    0,170 kg
  • W×D×H:
    71×26×142 mm
  • mV accuracy:
    ±0,1% FS
  • mV range:
    −1999...+2000; −1792...+2206 mVH (acc. to DIN 38404)
  • mV resolution:
    1
  • pH accuracy:
    ±0,01
  • pH calibration:
    1-, 2- or 3- point
  • pH range:
    0,00 - 14,00
  • pH resolution:
    0,01
  • Pk:
    1

Specifications

About this item

This meter is a simple handheld instrument for measuring pH, mV, temperature and relative humidity (0,0 to 70,0).

  • Battery operation makes it ideal for field measurements
  • Min./Max. value and HOLD memory
  • Impact-resistant ABS plastic housing, membrane keyboard, transparent panel; front side with IP 65 rating
  • Integrated pop-up clip for table top or suspended use
  • Serial interface (3,5 mm jack) can be connected to RS232 or USB interface of a PC via electrically isolated interface converter GRS 3100, GRS 3105 or USB 3100 N (available separately)
  • ORP mode allows for automatic conversion to hydrogen system electrodes
  • Automatic or manual temperature compensation
  • Automatic buffer detection
Delivery: Supplied with battery and manual. Electrodes need to be ordered separately.