GC septa in blister packs
Supplier: Thermo Fisher Scientific
Certificates
About this item
Quality GC septa for all applications. The range is now packaged into blister packaging for enhanced product cleanliness.
- Ultra-clean blister packaging
- Long lifetime material
- Maximum operating temperatures up to 400 °C