Gas washing bottles, with head, BOLA
Clear borosilicate glass 3.3 bottle with FEP inlet tube.
- Chemical resistant
- Temperature resistant from −200 to +250 °C
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Clear borosilicate glass 3.3 bottle with FEP inlet tube.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.