Order Entry
Finland
ContactUsLinkComponent
Bandage, OneMed
  SELA212573
undefined
Bandage, OneMed
  SELA212573
  • Description:
    Mollelast Adhesive, viscose and polyamide
  • L×W:
    20 m× 60 mm
  • Pk:
    1
Self-adhesive, elastic both lengthwise, crosswise and diagonally. Especially suitable for fixation where full mobility is sought, e.g. over joints. 66% viscose and 34% polyamide.